Product Details
- SNP ID
-
rs201753653
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
12
- Location
-
Chr.1:36319792 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGAGAACGCCCCCAGCTCCAAGAAG[A/G]TCCCGGCTCCTGACAAAGTCCCCTC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EVA1B
PubMed Links
Gene Details
- Gene
- EVA1B
- Gene Name
- eva-1 homolog B (C. elegans)
There are no transcripts associated with this gene.
- Gene
- SH3D21
- Gene Name
- SH3 domain containing 21
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001162530.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
NP_001156002.1 |
NM_024676.4 |
1202 |
Missense Mutation |
ATC,GTC |
I266V |
NP_078952.4 |
XM_011542153.2 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_011540455.1 |
XM_017002340.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_016857829.1 |
XM_017002341.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_016857830.1 |
XM_017002342.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_016857831.1 |
XM_017002343.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_016857832.1 |
XM_017002344.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_016857833.1 |
XM_017002345.1 |
1202 |
Missense Mutation |
ATC,GTC |
I377V |
XP_016857834.1 |
XM_017002346.1 |
1202 |
Missense Mutation |
ATC,GTC |
I23V |
XP_016857835.1 |
View Full Product Details