Product Details

SNP ID
rs199693209
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:152812720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGTCCCACTGCCACAGACCCCAG[A/C]GCTCTGGCTGCTGCAGCCAGCCCTC
Phenotype
MIM: 612604 MIM: 612605
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LCE1B PubMed Links

Gene Details

Gene
LCE1B
Gene Name
late cornified envelope 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178349.1 750 Missense Mutation AGC,CGC S92R NP_848126.1
Gene
LCE1C
Gene Name
late cornified envelope 1C
There are no transcripts associated with this gene.

View Full Product Details