Product Details

SNP ID
rs199694056
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:160343457 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGAAGTCGGGGTCTCCTTCGCCTT[C/T]TGTCTTTCTGCGTCCTACTAGCAGG
Phenotype
MIM: 601924 MIM: 605254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 402 UTR 5 NP_001091868.1
NM_004371.3 402 UTR 5 NP_004362.2
Gene
NCSTN
Gene Name
nicastrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290184.1 402 UTR 5 NP_001277113.1
NM_001290186.1 402 Silent Mutation CTG,TTG L21L NP_001277115.1
NM_015331.2 402 Silent Mutation CTG,TTG L21L NP_056146.1
XM_005245053.4 402 Silent Mutation CTG,TTG L21L XP_005245110.1
XM_011509363.2 402 Silent Mutation CTG,TTG L21L XP_011507665.1

View Full Product Details