Product Details

SNP ID
rs199921829
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:109250301 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCTACACCAGCCGCTGCAGGGA[G/T]GCGGGCACTGAGCTGACTGGCCACC
Phenotype
MIM: 604265
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CELSR2 PubMed Links

Gene Details

Gene
CELSR2
Gene Name
cadherin EGF LAG seven-pass G-type receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001408.2 283 Missense Mutation GAG,GAT E74D NP_001399.1
XM_005270580.4 283 Missense Mutation GAG,GAT E74D XP_005270637.1

View Full Product Details