Product Details

SNP ID
rs199523548
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:161984123 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGGCCTCCTCGTAGGCCACGTCA[C/T]GCAGCATGGCCCAGGCAGCCACGTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OLFML2B PubMed Links

Gene Details

Gene
OLFML2B
Gene Name
olfactomedin like 2B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297713.1 2846 Missense Mutation CAT,CGT H603R NP_001284642.1
NM_015441.2 2846 Missense Mutation CAT,CGT H602R NP_056256.1
XM_005245075.2 2846 Missense Mutation CAT,CGT H604R XP_005245132.1
XM_011509398.2 2846 Missense Mutation CAT,CGT H362R XP_011507700.1
XM_017000967.1 2846 Intron XP_016856456.1

View Full Product Details