Product Details

SNP ID
rs199708789
Assay Type
Functionally tested
NCBI dbSNP Submissions
4
Location
Chr.1:181089161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACCCGCCGGCTGGGGAGAGCCG[C/T]CCCCGCCCGCCGCTCGTGCCTCTTG
Phenotype
MIM: 607177
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IER5 PubMed Links

Gene Details

Gene
IER5
Gene Name
immediate early response 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016545.4 660 Missense Mutation CCC,TCC P87S NP_057629.2

View Full Product Details