Product Details
- SNP ID
-
rs199684312
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
9
- Location
-
Chr.1:18481360 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGAAAAGGCTCAGGTGAGGAGCGG[G/T]GCGGGCAGGGCTCGGACTCTGAGCA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KLHDC7A
PubMed Links
Gene Details
- Gene
- KLHDC7A
- Gene Name
- kelch domain containing 7A
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152375.2 |
431 |
Missense Mutation |
GGC,TGC |
G127C |
NP_689588.2 |
View Full Product Details