Product Details

SNP ID
rs200084732
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:53227045 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCATGTCCACACCAATATTCAGT[C/T]TAGATTGGTTTAATCTTGAAGTGTA
Phenotype
MIM: 602603
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf123 PubMed Links

Gene Details

Gene
C1orf123
Gene Name
chromosome 1 open reading frame 123
There are no transcripts associated with this gene.

Gene
MAGOH
Gene Name
mago homolog, exon junction complex core component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002370.3 603 Silent Mutation TAA,TAG *147* NP_002361.1

View Full Product Details