Product Details

SNP ID
rs199685043
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:6462065 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGAGCGCCTCCTGGGTCTCGGGG[C/T]AGCCAGGGGTCCAGCTGTTACCCAC
Phenotype
MIM: 606351 MIM: 611101 MIM: 603366
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ESPN PubMed Links

Gene Details

Gene
ESPN
Gene Name
espin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031475.2 831 Intron NP_113663.2
XM_005263501.2 831 Intron XP_005263558.1
XM_011542231.1 831 Intron XP_011540533.1
XM_011542232.1 831 Intron XP_011540534.1
XM_011542233.2 831 Intron XP_011540535.1
XM_011542236.2 831 Intron XP_011540538.1
XM_011542237.1 831 Intron XP_011540539.1
XM_011542238.2 831 Intron XP_011540540.1
XM_017002433.1 831 Intron XP_016857922.1
XM_017002434.1 831 Intron XP_016857923.1
Gene
PLEKHG5
Gene Name
pleckstrin homology and RhoGEF domain containing G5
There are no transcripts associated with this gene.

Gene
TNFRSF25
Gene Name
TNF receptor superfamily member 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039664.1 831 Intron NP_001034753.1
NM_003790.2 831 Missense Mutation TAC,TGC Y285C NP_003781.1
NM_148965.1 831 Missense Mutation TAC,TGC Y294C NP_683866.1
NM_148966.1 831 Missense Mutation TAC,TGC Y248C NP_683867.1
NM_148967.1 831 Missense Mutation TAC,TGC Y240C NP_683868.1
NM_148970.1 831 Missense Mutation TAC,TGC Y102C NP_683871.1

View Full Product Details