Product Details
- SNP ID
-
rs199954432
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
3
- Location
-
Chr.1:156037105 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGTTGAGCTGCTCCAGCTGCTGC[C/T]GAAATCTCACTTCTGGCGTCTGCAC
- Phenotype
-
MIM: 605440
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBQLN4
PubMed Links
Gene Details
- Gene
- UBQLN4
- Gene Name
- ubiquilin 4
View Full Product Details