Product Details
- SNP ID
-
rs200367748
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
1
- Location
-
Chr.1:182575436 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AACCCACTGGCCCCACCAGCTCCAT[C/T]ACACTGAGGCTTGTTTGGACTGTGG
- Phenotype
-
MIM: 180435
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
RNASEL
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs36053738] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RNASEL
- Gene Name
- ribonuclease L
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021133.3 |
2439 |
Missense Mutation |
AAT,GAT |
N728D |
NP_066956.1 |
View Full Product Details