Product Details

SNP ID
rs200923903
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:46248536 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAGGAGCTTGGCTCCCAGCCAG[C/G]TGGCCAAGAGAAAACCTGAAGGCAG
Phenotype
MIM: 603615
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RAD54L PubMed Links

Gene Details

Gene
RAD54L
Gene Name
RAD54-like (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142548.1 77 Missense Mutation CTG,GTG L10V NP_001136020.1
NM_003579.3 77 Missense Mutation CTG,GTG L10V NP_003570.2
XM_006710975.3 77 UTR 5 XP_006711038.1
XM_011542299.2 77 Intron XP_011540601.1
XM_011542300.2 77 Intron XP_011540602.1

View Full Product Details