Product Details

SNP ID
rs200402727
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:240493048 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCTGGATTTTCTTGAGTCGGAAG[A/G]GTGGGTCCAGGCCGGGGCACTCGAG
Phenotype
MIM: 608832
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GREM2 PubMed Links

Gene Details

Gene
GREM2
Gene Name
gremlin 2, DAN family BMP antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022469.3 708 Missense Mutation CCC,CTC P143L NP_071914.3
XM_005273226.3 708 Missense Mutation CCC,CTC P161L XP_005273283.1
XM_011544249.2 708 Missense Mutation CCC,CTC P143L XP_011542551.1

View Full Product Details