Product Details

SNP ID
rs200548746
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:41481119 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCAGCCTTTGGAGAAGCTCTCCT[A/G]TAGTGGCCCCTGTCTTGCCAGTCTG
Phenotype
MIM: 131241
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EDN2 PubMed Links

Gene Details

Gene
EDN2
Gene Name
endothelin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302269.1 501 Intron NP_001289198.1
NM_001956.4 501 Missense Mutation ACA,ATA T140I NP_001947.1
XM_017000512.1 501 Intron XP_016856001.1

View Full Product Details