Product Details

SNP ID
rs202206935
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:43338654 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGCGTCTCTTCTTTCCGCTGCAC[C/T]TCTGGGTGAAGAATGTGTTCCTAAA
Phenotype
MIM: 159530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MPL PubMed Links

Gene Details

Gene
MPL
Gene Name
MPL proto-oncogene, thrombopoietin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005373.2 370 Missense Mutation CTC,TTC L109F NP_005364.1
XM_017001320.1 370 Missense Mutation CTC,TTC L166F XP_016856809.1

View Full Product Details