Product Details

SNP ID
rs201864262
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:160343137 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTTTTATTCTCCACTACCTTTGA[C/T]CCGCGCGCTCTTGGTCTCGAATTTG
Phenotype
MIM: 601924 MIM: 605254
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COPA PubMed Links

Gene Details

Gene
COPA
Gene Name
coatomer protein complex subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098398.1 82 Missense Mutation ATC,GTC I12V NP_001091868.1
NM_004371.3 82 Missense Mutation ATC,GTC I12V NP_004362.2
Gene
NCSTN
Gene Name
nicastrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290184.1 82 Intron NP_001277113.1
NM_001290186.1 82 Intron NP_001277115.1
NM_015331.2 82 Intron NP_056146.1
XM_005245053.4 82 UTR 5 XP_005245110.1
XM_011509363.2 82 UTR 5 XP_011507665.1

View Full Product Details