Product Details

SNP ID
rs201983292
Assay Type
Functionally Tested
NCBI dbSNP Submissions
9
Location
Chr.1:220696691 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTTCGCTGCAGCCTACTCCGCCC[C/T]GTTTGCGGTAGCCACCAGCGTGGTA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C1orf115 PubMed Links

Gene Details

Gene
C1orf115
Gene Name
chromosome 1 open reading frame 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024709.4 506 Missense Mutation CCG,CTG P130L NP_078985.3

View Full Product Details