Product Details

SNP ID
rs201996831
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:16015625 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGGTCTGCTGGACGTGTTCTGT[A/G]TGCGGGTGACGCCGTGCCCGGATAG
Phenotype
MIM: 602024 MIM: 610692
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C1orf64 PubMed Links

Gene Details

Gene
C1orf64
Gene Name
chromosome 1 open reading frame 64
There are no transcripts associated with this gene.

Gene
CLCNKA
Gene Name
chloride voltage-gated channel Ka
There are no transcripts associated with this gene.

Gene
HSPB7
Gene Name
heat shock protein family B (small) member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014424.4 1295 Silent Mutation CAC,CAT H156H NP_055239.1
XM_011541249.1 1295 Silent Mutation CAC,CAT H249H XP_011539551.1
XM_011541250.2 1295 Silent Mutation CAC,CAT H248H XP_011539552.1

View Full Product Details