Product Details

SNP ID
rs202012464
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:110517369 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAATTTCTCCTGGGATGTTCTG[C/T]TTTTCTTCATTCTCAGTCTCCCGGT
Phenotype
MIM: 602420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNA10 PubMed Links

Gene Details

Gene
KCNA10
Gene Name
potassium voltage-gated channel subfamily A member 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005549.2 1807 Silent Mutation AAA,AAG K473K NP_005540.1

View Full Product Details