Product Details

SNP ID
rs201978159
Assay Type
Functionally tested
NCBI dbSNP Submissions
3
Location
Chr.1:32826184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTGCCCCATTTTCTTGGGATTCC[A/T]TGGATGAGGATGGATTGGATGACTC
Phenotype
MIM: 611889 MIM: 603623
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
S100PBP PubMed Links

Gene Details

Gene
S100PBP
Gene Name
S100P binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256121.1 311 Missense Mutation ATG,TTG M29L NP_001243050.1
NM_022753.3 311 Missense Mutation ATG,TTG M29L NP_073590.2
XM_011541961.2 311 Missense Mutation ATG,TTG M29L XP_011540263.1
XM_011541962.2 311 Missense Mutation ATG,TTG M29L XP_011540264.1
XM_011541963.1 311 Missense Mutation ATG,TTG M29L XP_011540265.1
XM_017002098.1 311 Missense Mutation ATG,TTG M29L XP_016857587.1
Gene
YARS
Gene Name
tyrosyl-tRNA synthetase
There are no transcripts associated with this gene.

View Full Product Details