Product Details

SNP ID
rs202032042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:2865415 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTGCAGGCTCTGGTGGCTGAAG[A/C]TGACTGCCCTGGCAGATTTGGAAGA
Phenotype
MIM: 176884 MIM: 608550
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PTPRA PubMed Links

Gene Details

Gene
PTPRA
Gene Name
protein tyrosine phosphatase, receptor type A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002836.3 2269 Intron NP_002827.1
NM_080840.2 2269 Intron NP_543030.1
NM_080841.2 2269 Intron NP_543031.1
Gene
VPS16
Gene Name
VPS16, CORVET/HOPS core subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022575.3 2269 Missense Mutation ATG,CTG M731L NP_072097.2
NM_080413.2 2269 Missense Mutation ATG,CTG M587L NP_536338.1

View Full Product Details