Product Details

SNP ID
rs200241491
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:58997639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTGCATAGATTTCTGCCATCATC[A/T]TCTCCCTCCCAGAGAGGGAGCCGTA
Phenotype
MIM: 603169 MIM: 605297
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CTSZ PubMed Links

Gene Details

Gene
CTSZ
Gene Name
cathepsin Z
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001336.3 728 Missense Mutation AAG,ATG K201M NP_001327.2
Gene
NELFCD
Gene Name
negative elongation factor complex member C/D
There are no transcripts associated with this gene.

View Full Product Details