Product Details

SNP ID
rs200152114
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57561604 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGCCAGATGGAGGAAGAGGGC[A/G]TCCTCAGGCGGCTGAAGAAGTATGA
Phenotype
MIM: 614168
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCK1 PubMed Links

Gene Details

Gene
PCK1
Gene Name
phosphoenolpyruvate carboxykinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002591.3 356 Missense Mutation ATC,GTC I65V NP_002582.3
XM_011528839.1 356 UTR 5 XP_011527141.1

View Full Product Details