Product Details

SNP ID
rs199753286
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:19479979 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGCCGCCGTGGCTATGTTCGTGT[A/C]CGATTTCCGCAAAGAGTTCTACGAG
Phenotype
MIM: 603465 MIM: 601754
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDC45 PubMed Links

Gene Details

Gene
CDC45
Gene Name
cell division cycle 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001178010.2 154 Missense Mutation TAC,TCC Y4S NP_001171481.1
NM_001178011.2 154 Missense Mutation TAC,TCC Y4S NP_001171482.1
NM_003504.4 154 Missense Mutation TAC,TCC Y4S NP_003495.1
XM_005261285.2 154 Missense Mutation TAC,TCC Y4S XP_005261342.1
XM_005261286.3 154 Missense Mutation TAC,TCC Y4S XP_005261343.1
XM_011530415.1 154 Missense Mutation TAC,TCC Y4S XP_011528717.1
XM_011530416.1 154 Intron XP_011528718.1
XM_011530417.2 154 Missense Mutation TAC,TCC Y4S XP_011528719.1
XM_011530418.2 154 Missense Mutation TAC,TCC Y4S XP_011528720.1
XM_017028966.1 154 Intron XP_016884455.1
XM_017028967.1 154 Intron XP_016884456.1
Gene
UFD1L
Gene Name
ubiquitin fusion degradation 1 like (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035247.2 154 Intron NP_001030324.2
NM_005659.6 154 Intron NP_005650.2
XM_011530361.2 154 Intron XP_011528663.1

View Full Product Details