Product Details
- SNP ID
-
rs201620023
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:49962855 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGCCGCCTGCTCTTCCGGAGGAG[A/G]GTCTCTCCAGGTGCGTGGTGGCTCG
- Phenotype
-
MIM: 610580
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR6821
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4078443] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR6821
- Gene Name
- microRNA 6821
There are no transcripts associated with this gene.
- Gene
- PIM3
- Gene Name
- Pim-3 proto-oncogene, serine/threonine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001001852.3 |
1236 |
Silent Mutation |
AGA,AGG |
R261R |
NP_001001852.2 |
View Full Product Details