Product Details
- SNP ID
-
rs201677987
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:31768846 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CATGTGCCTATATCACCCAGAAGGT[C/G]GAGTTTGCTGGCATCAGGTAGATAT
- Phenotype
-
MIM: 614191
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DEPDC5
PubMed Links
Gene Details
- Gene
- DEPDC5
- Gene Name
- DEP domain containing 5
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001007188.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
NP_001007189.1 |
NM_001136029.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
NP_001129501.1 |
NM_001242896.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
NP_001229825.1 |
NM_001242897.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
NP_001229826.1 |
NM_014662.4 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
NP_055477.1 |
XM_005261862.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_005261919.1 |
XM_011530557.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528859.1 |
XM_011530558.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528860.1 |
XM_011530559.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528861.1 |
XM_011530560.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528862.1 |
XM_011530561.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528863.1 |
XM_011530562.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528864.1 |
XM_011530563.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528865.1 |
XM_011530565.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528867.1 |
XM_011530568.2 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_011528870.1 |
XM_011530569.2 |
502 |
Intron |
|
|
XP_011528871.1 |
XM_017029109.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884598.1 |
XM_017029110.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884599.1 |
XM_017029111.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884600.1 |
XM_017029112.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884601.1 |
XM_017029113.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884602.1 |
XM_017029114.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884603.1 |
XM_017029115.1 |
502 |
Silent Mutation |
GTC,GTG |
V132V |
XP_016884604.1 |
View Full Product Details