Product Details
- SNP ID
-
rs201800009
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:23743968 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCTTTGCCACACTCGCAGGTGTA[A/G]GGCTTCTTGCCGGTGTGGATTCTCT
- Phenotype
-
MIM: 605017
MIM: 194544
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
VPREB3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs5759985] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- VPREB3
- Gene Name
- pre-B lymphocyte 3
There are no transcripts associated with this gene.
- Gene
- ZNF70
- Gene Name
- zinc finger protein 70
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021916.3 |
1634 |
Silent Mutation |
CCC,CCT |
P391P |
NP_068735.1 |
View Full Product Details