Product Details

SNP ID
rs199881041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:32479063 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACCTTGGCTTCATATGGGATT[A/G]TTTCTGGGGACTTGATATGTTTGAT
Phenotype
MIM: 605648
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FBXO7 PubMed Links

Gene Details

Gene
FBXO7
Gene Name
F-box protein 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033024.1 488 Intron NP_001028196.1
NM_001257990.1 488 UTR 5 NP_001244919.1
NM_012179.3 488 Missense Mutation ATT,GTT I69V NP_036311.3
XM_011530106.1 488 Intron XP_011528408.1

View Full Product Details