Product Details

SNP ID
rs200943463
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:79027811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCCTCCTCTTCTCTATAGGGCT[C/T]TGAGCCTGATGGAGATGGATGGGAG
Phenotype
MIM: 609933
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
REG3G PubMed Links

Gene Details

Gene
REG3G
Gene Name
regenerating family member 3 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008387.2 Intron NP_001008388.1
NM_001270040.1 Intron NP_001256969.1
NM_198448.3 Intron NP_940850.1
XM_005264135.2 Intron XP_005264192.1

View Full Product Details