Product Details

SNP ID
rs201293992
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:181678111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCAAAGAAGGGCTCCAGCGCTGC[G/T]CTGTAGGCGTGCGGCGGAGGCTTAA
Phenotype
MIM: 601724
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NEUROD1 PubMed Links

Gene Details

Gene
NEUROD1
Gene Name
neuronal differentiation 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002500.4 997 Silent Mutation NP_002491.2

View Full Product Details