Product Details

SNP ID
rs201436890
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:73240491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAATTCTGAAACTTCTTGATGTTG[C/T]CCATCCTGCAGCAAAGACTTTGGTA
Phenotype
MIM: 605140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT7 PubMed Links

Gene Details

Gene
CCT7
Gene Name
chaperonin containing TCP1 subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001009570.2 358 Intron NP_001009570.1
NM_001166284.1 358 Intron NP_001159756.1
NM_001166285.1 358 Missense Mutation GCC,GTC A28V NP_001159757.1
NM_006429.3 358 Missense Mutation GCC,GTC A72V NP_006420.1
XM_011532478.2 358 Missense Mutation GCC,GTC A28V XP_011530780.1
XM_011532479.1 358 Missense Mutation GCC,GTC A28V XP_011530781.1
Gene
PRADC1
Gene Name
protease associated domain containing 1
There are no transcripts associated with this gene.

View Full Product Details