Product Details

SNP ID
rs200080144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95149530 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGAAGGTTCTCCCACATTCCC[A/G]GCATTCATAGGGCTTTTCTCCAGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF514 PubMed Links

Gene Details

Gene
ZNF514
Gene Name
zinc finger protein 514
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318005.1 1776 Silent Mutation NP_001304934.1
NM_032788.2 1776 Silent Mutation NP_116177.1
XM_006712806.2 1776 Missense Mutation XP_006712869.1

View Full Product Details