Product Details

SNP ID
rs200569195
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70936037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCAGCCCGAGAACACATGCAGG[C/T]GGTCACCCGAAACTACATCACCCAC
Phenotype
MIM: 192132 MIM: 604295
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP6V1B1 PubMed Links

Gene Details

Gene
ATP6V1B1
Gene Name
ATPase H+ transporting V1 subunit B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001692.3 170 Missense Mutation GCG,GTG A28V NP_001683.2
XM_011532907.2 170 Intron XP_011531209.1
Gene
ATP6V1B1-AS1
Gene Name
ATP6V1B1 antisense RNA 1
There are no transcripts associated with this gene.

Gene
VAX2
Gene Name
ventral anterior homeobox 2
There are no transcripts associated with this gene.

View Full Product Details