Product Details

SNP ID
rs200206979
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTGACATTTTGCCTTAAGCGGTA[A/G]CGTCTATAAGCACGCTGAATGACAG
Phenotype
MIM: 603415
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 5981 Silent Mutation CGC,CGT R1891R NP_002968.1
XM_005246757.2 5981 Silent Mutation CGC,CGT R1902R XP_005246814.1
XM_011511616.2 5981 Silent Mutation CGC,CGT R1902R XP_011509918.1
XM_011511617.2 5981 Silent Mutation CGC,CGT R1902R XP_011509919.1
XM_011511618.2 5981 Silent Mutation CGC,CGT R1891R XP_011509920.1
XM_011511619.2 5981 Intron XP_011509921.1
XM_017004668.1 5981 Silent Mutation CGC,CGT R1773R XP_016860157.1
XM_017004669.1 5981 Silent Mutation CGC,CGT R1654R XP_016860158.1

View Full Product Details