Product Details

SNP ID
rs200439527
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74423910 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATCCCAGCAAAGGGTCCCAACAT[C/T]GTACTGAGCGAGGAGCTGGCTGGGC
Phenotype
MIM: 602288
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf81 PubMed Links

Gene Details

Gene
C2orf81
Gene Name
chromosome 2 open reading frame 81
There are no transcripts associated with this gene.

Gene
RTKN
Gene Name
rhotekin
There are no transcripts associated with this gene.

Gene
WDR54
Gene Name
WD repeat domain 54
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320823.1 336 Silent Mutation ATC,ATT I169I NP_001307752.1
NM_001320824.1 336 Silent Mutation ATC,ATT I154I NP_001307753.1
NM_001320825.1 336 Silent Mutation ATC,ATT I102I NP_001307754.1
NM_032118.3 336 Silent Mutation ATC,ATT I154I NP_115494.1
XM_017005064.1 336 Silent Mutation ATC,ATT I102I XP_016860553.1

View Full Product Details