Product Details

SNP ID
rs200523042
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27276815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGGATCTTAAGAAGGGCGCCTC[A/T]CCTGAAGACTTCAAAAAATCCTACA
Phenotype
MIM: 613946 MIM: 602878 MIM: 606474
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAJC5G PubMed Links

Gene Details

Gene
DNAJC5G
Gene Name
DnaJ heat shock protein family (Hsp40) member C5 gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303127.1 443 Silent Mutation TCA,TCT S29S NP_001290056.1
NM_001303128.1 443 Silent Mutation TCA,TCT S29S NP_001290057.1
NM_173650.2 443 Silent Mutation TCA,TCT S29S NP_775921.1
XM_006711995.3 443 Silent Mutation TCA,TCT S29S XP_006712058.1
XM_017003872.1 443 Silent Mutation TCA,TCT S29S XP_016859361.1
XM_017003873.1 443 Silent Mutation TCA,TCT S29S XP_016859362.1
XM_017003874.1 443 Silent Mutation TCA,TCT S29S XP_016859363.1
Gene
SLC30A3
Gene Name
solute carrier family 30 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318949.1 443 Intron NP_001305878.1
NM_001318950.1 443 Intron NP_001305879.1
NM_001318951.1 443 Intron NP_001305880.1
NM_003459.4 443 Intron NP_003450.2
XM_005264547.1 443 Intron XP_005264604.1
XM_005264548.3 443 Intron XP_005264605.1
XM_006712100.2 443 Intron XP_006712163.1
XM_011533102.2 443 Intron XP_011531404.1
XM_011533103.2 443 Intron XP_011531405.1
XM_017004873.1 443 Intron XP_016860362.1
XM_017004874.1 443 Intron XP_016860363.1
XM_017004875.1 443 Intron XP_016860364.1
Gene
TRIM54
Gene Name
tripartite motif containing 54
There are no transcripts associated with this gene.

View Full Product Details