Product Details

SNP ID
rs202170773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96144222 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTGGTTGTCCTCCACAGGGATA[C/T]TCTTGTAGCGGAAAAGGCCCTCAAA
Phenotype
MIM: 608860 MIM: 603068
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASTL PubMed Links

Gene Details

Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
There are no transcripts associated with this gene.

Gene
DUSP2
Gene Name
dual specificity phosphatase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004418.3 748 Missense Mutation NP_004409.1
XM_017003546.1 748 Missense Mutation XP_016859035.1

View Full Product Details