Product Details
- SNP ID
-
rs201629421
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:100569717 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGGTCATCTTGCACCTTTACAAA[C/T]AAGGGTGAGCTGATCTTCCACTCCA
- Phenotype
-
MIM: 611678
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PDCL3
PubMed Links
Gene Details
- Gene
- PDCL3
- Gene Name
- phosducin like 3
View Full Product Details