Product Details

SNP ID
rs201656083
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:200818327 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATGTTCGGAATCAGCAAACCAT[C/T]GGAGCTCGTAAGGAGCTCCAGAAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BZW1 PubMed Links

Gene Details

Gene
BZW1
Gene Name
basic leucine zipper and W2 domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207067.1 1208 Silent Mutation ATC,ATT I251I NP_001193996.1
NM_001207069.1 1208 Intron NP_001193998.1
NM_001321688.1 1208 Intron NP_001308617.1
NM_001321690.1 1208 Intron NP_001308619.1
NM_001321691.1 1208 Intron NP_001308620.1
NM_001321693.1 1208 Intron NP_001308622.1
NM_001321694.1 1208 Intron NP_001308623.1
NM_014670.3 1208 Silent Mutation ATC,ATT I251I NP_055485.2
XM_017005393.1 1208 Intron XP_016860882.1
Gene
LOC101927795
Gene Name
uncharacterized LOC101927795
There are no transcripts associated with this gene.

View Full Product Details