Product Details

SNP ID
rs201893095
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:176109072 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGCAAGTCAAAATCTGGTTCCAGA[A/G]TCGCAGGATGAAAGAAAAGAAACTG
Phenotype
MIM: 142984 MIM: 142986 MIM: 142988
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HOXD10 PubMed Links

Gene Details

Gene
HOXD10
Gene Name
homeobox D10
There are no transcripts associated with this gene.

Gene
HOXD11
Gene Name
homeobox D11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021192.2 947 Missense Mutation AAT,AGT N316S NP_067015.2
Gene
HOXD12
Gene Name
homeobox D12
There are no transcripts associated with this gene.

Gene
LOC107985805
Gene Name
sine oculis-binding protein homolog
There are no transcripts associated with this gene.

View Full Product Details