Product Details

SNP ID
rs201972402
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:9948847 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGATGAGAGACTTGGGGGTGCCT[G/T]GGGCCTCCACAGCTCCTCTCACCCG
Phenotype
MIM: 607170
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRELD1 PubMed Links

Gene Details

Gene
CRELD1
Gene Name
cysteine rich with EGF like domains 1
There are no transcripts associated with this gene.

Gene
PRRT3
Gene Name
proline rich transmembrane protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318871.1 3098 Missense Mutation CAA,CCA Q361P NP_001305800.1
NM_207351.4 3098 Missense Mutation CAA,CCA Q361P NP_997234.3
XM_011533628.2 3098 Missense Mutation CAA,CCA Q361P XP_011531930.1
XM_011533629.2 3098 Missense Mutation CAA,CCA Q361P XP_011531931.1
XM_017006256.1 3098 Missense Mutation CAA,CCA Q361P XP_016861745.1
Gene
PRRT3-AS1
Gene Name
PRRT3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details