Product Details

SNP ID
rs201193706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44354807 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGTTTTGCCACCTGAGACCTATG[A/G]GGAGGTAAGCATCATGGTCCAATCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCAIM PubMed Links

Gene Details

Gene
TCAIM
Gene Name
T-cell activation inhibitor, mitochondrial
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029839.2 452 Missense Mutation AGG,GGG R9G NP_001025010.1
NM_001029840.2 452 Missense Mutation AGG,GGG R9G NP_001025011.1
NM_001282913.1 452 Missense Mutation AGG,GGG R9G NP_001269842.1
NM_001282914.1 452 UTR 5 NP_001269843.1
NM_001282915.1 452 UTR 5 NP_001269844.1
NM_173826.3 452 Missense Mutation AGG,GGG R9G NP_776187.2
XM_017006251.1 452 UTR 5 XP_016861740.1

View Full Product Details