Product Details

SNP ID
rs201331498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:44629325 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTACCATGAGACATCTGGACCCCA[A/G]GAAGCCCTGAGCCGGCTCAGGGAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF197 PubMed Links

Gene Details

Gene
ZNF197
Gene Name
zinc finger protein 197
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005264783.4 360 Silent Mutation CAA,CAG Q57Q XP_005264840.1
XM_017005495.1 360 Silent Mutation CAA,CAG Q57Q XP_016860984.1
Gene
ZNF197-AS1
Gene Name
ZNF197 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details