Product Details

SNP ID
rs200875272
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:125453823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCATCTTTACCTGCCTTCCAGAT[G/T]TTTAGACTTTAGCTGTTGTTCTCCT
Phenotype
MIM: 605931
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SNX4 PubMed Links

Gene Details

Gene
SNX4
Gene Name
sorting nexin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003794.3 1307 Missense Mutation AAT,CAT N393H NP_003785.1
XM_017007414.1 1307 Missense Mutation AAT,CAT N425H XP_016862903.1

View Full Product Details