Product Details

SNP ID
rs201104682
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:143266892 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAATTTATGGCTAGTTCATCCTGG[C/T]TTACAATGCATTCCACATCATCCTC
Phenotype
MIM: 608396
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC9A9 PubMed Links

Gene Details

Gene
SLC9A9
Gene Name
solute carrier family 9 member A9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173653.3 1957 Missense Mutation AAC,AGC N583S NP_775924.1
XM_011512703.2 1957 Missense Mutation AAC,AGC N367S XP_011511005.1
XM_011512704.2 1957 Intron XP_011511006.1
XM_017006202.1 1957 Intron XP_016861691.1
XM_017006203.1 1957 Missense Mutation AAC,AGC N466S XP_016861692.1

View Full Product Details