Product Details

SNP ID
rs201313857
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42908837 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGCTCTGATTTCCCAGCTGGAG[C/T]GAGGGGAAACACCCTGGTGCTCGGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF662 PubMed Links

Gene Details

Gene
ZNF662
Gene Name
zinc finger protein 662
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134656.1 445 Nonsense Mutation CGA,TGA R87* NP_001128128.1
NM_207404.3 445 Nonsense Mutation CGA,TGA R27* NP_997287.2

View Full Product Details