Product Details

SNP ID
rs200165076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:108908111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGCTGCTTCCCATTACAGATTA[C/T]TCTCAGCACATTGGAGAAGTCGAAG
Phenotype
MIM: 605128
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GUCA1C PubMed Links

Gene Details

Gene
GUCA1C
Gene Name
guanylate cyclase activator 1C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005459.3 609 Missense Mutation ATA,GTA I181V NP_005450.3
XM_005247910.3 609 Missense Mutation AAT,AGT N194S XP_005247967.1
XM_011513334.2 609 Missense Mutation ATA,GTA I97V XP_011511636.1

View Full Product Details