Product Details

SNP ID
rs200527724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:69025594 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGATTTCCCCTTCCCTTAGCTTT[A/G]GCTGAGACTGTAGGTTTTCAATTAT
Phenotype
MIM: 601126
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMF1 PubMed Links

Gene Details

Gene
TMF1
Gene Name
TATA element modulatory factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007114.2 3225 Missense Mutation CCA,CTA P993L NP_009045.2
XM_011534056.2 3225 Missense Mutation CCA,CTA P996L XP_011532358.1

View Full Product Details