Product Details
- SNP ID
-
rs200304933
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:52548109 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTTGGCGAATGTTGAGGGTGTCC[C/T]GGAGCATCAAATCTCGAAGGCGCCA
- Phenotype
-
MIM: 606083
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PBRM1
PubMed Links
Gene Details
- Gene
- PBRM1
- Gene Name
- polybromo 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018313.4 |
5228 |
Missense Mutation |
CAG,CGG |
Q1568R |
NP_060783.3 |
XM_005265279.4 |
5228 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_005265336.1 |
XM_005265280.3 |
5228 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_005265337.1 |
XM_005265282.3 |
5228 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_005265339.1 |
XM_005265283.3 |
5228 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_005265340.1 |
XM_011533900.2 |
5228 |
Missense Mutation |
CAG,CGG |
Q1711R |
XP_011532202.1 |
XM_011533902.2 |
5228 |
Missense Mutation |
CAG,CGG |
Q1711R |
XP_011532204.1 |
XM_011533903.2 |
5228 |
Missense Mutation |
CAG,CGG |
Q1711R |
XP_011532205.1 |
XM_017006725.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1720R |
XP_016862214.1 |
XM_017006726.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1720R |
XP_016862215.1 |
XM_017006727.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1720R |
XP_016862216.1 |
XM_017006728.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1720R |
XP_016862217.1 |
XM_017006729.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1719R |
XP_016862218.1 |
XM_017006730.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1717R |
XP_016862219.1 |
XM_017006731.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1717R |
XP_016862220.1 |
XM_017006732.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1710R |
XP_016862221.1 |
XM_017006733.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1707R |
XP_016862222.1 |
XM_017006734.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1707R |
XP_016862223.1 |
XM_017006735.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1705R |
XP_016862224.1 |
XM_017006736.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1704R |
XP_016862225.1 |
XM_017006737.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1699R |
XP_016862226.1 |
XM_017006738.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1699R |
XP_016862227.1 |
XM_017006739.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1699R |
XP_016862228.1 |
XM_017006740.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1699R |
XP_016862229.1 |
XM_017006741.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1696R |
XP_016862230.1 |
XM_017006742.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1696R |
XP_016862231.1 |
XM_017006743.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1695R |
XP_016862232.1 |
XM_017006744.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1693R |
XP_016862233.1 |
XM_017006745.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_016862234.1 |
XM_017006746.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1690R |
XP_016862235.1 |
XM_017006747.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1688R |
XP_016862236.1 |
XM_017006748.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1675R |
XP_016862237.1 |
XM_017006749.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1675R |
XP_016862238.1 |
XM_017006750.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1675R |
XP_016862239.1 |
XM_017006751.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1671R |
XP_016862240.1 |
XM_017006752.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1668R |
XP_016862241.1 |
XM_017006753.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1665R |
XP_016862242.1 |
XM_017006754.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1659R |
XP_016862243.1 |
XM_017006755.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1641R |
XP_016862244.1 |
XM_017006756.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1640R |
XP_016862245.1 |
XM_017006757.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1620R |
XP_016862246.1 |
XM_017006758.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1620R |
XP_016862247.1 |
XM_017006759.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1616R |
XP_016862248.1 |
XM_017006760.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1604R |
XP_016862249.1 |
XM_017006761.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1604R |
XP_016862250.1 |
XM_017006762.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1603R |
XP_016862251.1 |
XM_017006763.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1601R |
XP_016862252.1 |
XM_017006764.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1589R |
XP_016862253.1 |
XM_017006765.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1568R |
XP_016862254.1 |
XM_017006766.1 |
5228 |
Missense Mutation |
CAG,CGG |
Q1564R |
XP_016862255.1 |
- Gene
- SMIM4
- Gene Name
- small integral membrane protein 4
There are no transcripts associated with this gene.
View Full Product Details