Product Details

SNP ID
rs199621885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:191461291 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAAAGGAGCTACAGACCGTCCC[C/T]CAGACAGAGGTAGACAAGGCTAATG
Phenotype
MIM: 615701
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINCR-0002 PubMed Links

Gene Details

Gene
LINCR-0002
Gene Name
uncharacterized LincR-0002
There are no transcripts associated with this gene.

Gene
PYDC2
Gene Name
pyrin domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083308.1 Intron NP_001076777.1

View Full Product Details